Assay Details
| Gene Symbol: sec-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069743.6 | NP_502144.1 | 12 - 13 | 2609 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sec-31| Gene Symbol: sec-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069743.6 | NP_502144.1 | 12 - 13 | 2609 | 94 | |
Set Membership: |
|