Assay Details
| Gene Symbol: magi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268648.3 | NP_001255577.1 | 2 - 3 | 137 | 106 | |
| NM_001383205.1 | 2 - 3 | 127 | 106 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

magi-1| Gene Symbol: magi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268648.3 | NP_001255577.1 | 2 - 3 | 137 | 106 | |
| NM_001383205.1 | 2 - 3 | 127 | 106 | |||
Set Membership: |
|