Assay Details
| Gene Symbol: magi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268648.3 | NP_001255577.1 | 9 - 10 | 1132 | 98 | |
| NM_001268650.4 | NP_001255579.1 | 4 - 5 | 722 | 98 | ||
| NM_001268652.3 | NP_001255581.1 | 2 - 3 | 406 | 98 | ||
| NM_001383205.1 | 9 - 10 | 1122 | 98 | |||
| NM_001383206.1 | 9 - 10 | 1138 | 98 | |||
More Information
Set Membership: |
|

magi-1
Genomic Map