Assay Details
| Gene Symbol: magi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268648.3 | NP_001255577.1 | 11 - 12 | 2061 | 67 | |
| NM_001268650.4 | NP_001255579.1 | 6 - 7 | 1651 | 67 | ||
| NM_001268652.3 | NP_001255581.1 | 4 - 5 | 1335 | 67 | ||
| NM_001383205.1 | 11 - 12 | 2051 | 67 | |||
| NM_001383206.1 | 11 - 12 | 2067 | 67 | |||
More Information
Set Membership: |
|

magi-1
Genomic Map