Assay Details
| Gene Symbol: magi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268648.3 | NP_001255577.1 | 12 - 13 | 2948 | 91 | |
| NM_001268650.4 | NP_001255579.1 | 7 - 8 | 2538 | 91 | ||
| NM_001268652.3 | NP_001255581.1 | 5 - 6 | 2222 | 91 | ||
| NM_001383206.1 | 12 - 13 | 2954 | 91 | |||
More Information
Set Membership: |
|

magi-1
Genomic Map