Assay Details
| Gene Symbol: ddb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069898.8 | NP_502299.1 | 10 - 11 | 3307 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddb-1| Gene Symbol: ddb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069898.8 | NP_502299.1 | 10 - 11 | 3307 | 118 | |
Set Membership: |
|