Assay Details
| Gene Symbol: mlt-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069918.4 | NP_502319.2 | 4 - 5 | 1054 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlt-3| Gene Symbol: mlt-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069918.4 | NP_502319.2 | 4 - 5 | 1054 | 76 | |
Set Membership: |
|