Assay Details
| Gene Symbol: vps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027888.5 | NP_001023059.1 | 11 - 12 | 3609 | 68 | |
| NM_001307062.3 | NP_001293991.1 | 9 - 10 | 3189 | 68 | ||
| NM_001307063.3 | NP_001293992.1 | 1 - 2 | 90 | 68 | ||
| NM_001372932.2 | NP_001359706.1 | 3 - 4 | 383 | 68 | ||
More Information
Set Membership: |
|

vps-8
Genomic Map