Assay Details
| Gene Symbol: vps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027888.5 | NP_001023059.1 | 6 - 7 | 2246 | 125 | |
| NM_001307062.3 | NP_001293991.1 | 4 - 5 | 1826 | 125 | ||
More Information
Set Membership: |
|

vps-8
Genomic Map