Assay Details
| Gene Symbol: dpy-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069977.8 | NP_502378.1 | 2 - 3 | 430 | 124 | |
| GenBank mRNA | U43562.1 | - | 2 - 3 | 385 | 124 | |
More Information
Set Membership: |
|

dpy-26
Genomic Map