Assay Details
| Gene Symbol: prmt-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047525.5 | NP_001040990.1 | 9 - 10 | 1979 | 90 | |
| NM_001047526.3 | NP_001040991.1 | 5 - 6 | 980 | 90 | ||
More Information
Set Membership: |
|

prmt-9
Genomic Map