Assay Details
| Gene Symbol: C27D8.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070024.7 | NP_502425.2 | 5 - 6 | 747 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C27D8.4| Gene Symbol: C27D8.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070024.7 | NP_502425.2 | 5 - 6 | 747 | 112 | |
Set Membership: |
|