Assay Details
Gene Symbol: inx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070034.5 | NP_502435.1 | 2 - 3 | 240 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
