Assay Details
| Gene Symbol: gcy-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070048.7 | NP_502449.2 | 8 - 9 | 1422 | 155 | |
| GenBank mRNA | AB201389.1 | - | 8 - 9 | 1382 | 155 | |
More Information
Set Membership: |
|

gcy-18
Genomic Map