Assay Details
Gene Symbol: dod-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268760.2 | NP_001255689.1 | 2 - 3 | 355 | 118 | |
NM_001268761.3 | NP_001255690.1 | 1 - 2 | 152 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
