Assay Details
Gene Symbol: spe-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268773.1 | NP_001255702.1 | 3 - 4 | 285 | 64 | |
NM_001268774.1 | NP_001255703.1 | 3 - 4 | 285 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
