Assay Details
Gene Symbol: lin-54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372945.2 | NP_001360500.1 | 5 - 6 | 903 | 94 | |
NM_001372946.3 | NP_001360501.1 | 5 - 6 | 903 | 94 | ||
NM_070143.4 | NP_502544.1 | 5 - 6 | 906 | 94 | ||
NM_070144.6 | NP_502545.1 | 5 - 6 | 895 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
