Assay Details
Gene Symbol: lin-54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372946.3 | NP_001360501.1 | 1 - 2 | 148 | 79 | |
NM_070144.6 | NP_502545.1 | 1 - 2 | 149 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
