Assay Details
Gene Symbol: lin-54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372945.2 | NP_001360500.1 | 2 - 3 | 302 | 87 | |
NM_070144.6 | NP_502545.1 | 2 - 3 | 294 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
