Assay Details
| Gene Symbol: fat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070159.10 | NP_502560.1 | 2 - 3 | 798 | 106 | |
| GenBank mRNA | AF240777.1 | - | 2 - 3 | 796 | 106 | |
More Information
Set Membership: |
|

fat-2
Genomic Map