Assay Details
| Gene Symbol: paqr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306938.3 | NP_001293867.1 | 3 - 4 | 441 | 127 | |
| NM_070344.7 | NP_502745.2 | 4 - 5 | 581 | 127 | ||
More Information
Set Membership: |
|

paqr-3
Genomic Map