Assay Details
| Gene Symbol: eps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047582.5 | NP_001041047.1 | 1 - 2 | 127 | 90 | |
| NM_001047583.7 | NP_001041048.1 | 1 - 2 | 133 | 90 | ||
| NM_001306998.2 | NP_001293927.1 | 1 - 2 | 126 | 90 | ||
| NM_001372963.2 | NP_001360610.1 | 1 | 54 | 90 | ||
| NM_001372964.2 | NP_001360611.1 | 1 | 54 | 90 | ||
| GenBank mRNA | AY611484.1 | - | 1 - 2 | 51 | 90 | |
| AY611485.1 | - | 1 - 2 | 51 | 90 | ||
More Information
Set Membership: |
|

eps-8
Genomic Map