Assay Details
| Gene Symbol: qars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268914.4 | NP_001255843.1 | 5 - 6 | 2243 | 124 | |
| NM_001268915.3 | NP_001255844.1 | 3 - 4 | 1558 | 124 | ||
| NM_001268916.3 | NP_001255845.1 | 2 - 3 | 1126 | 124 | ||
| NM_001276892.4 | NP_001263821.1 | 1 - 2 | 514 | 124 | ||
More Information
Set Membership: |
|

qars-1
Genomic Map