Assay Details
| Gene Symbol: cpna-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070453.5 | NP_502854.2 | 8 - 9 | 1304 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpna-4| Gene Symbol: cpna-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070453.5 | NP_502854.2 | 8 - 9 | 1304 | 74 | |
Set Membership: |
|