Assay Details
Gene Symbol: ntl-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307177.4 | NP_001294106.1 | 4 - 5 | 970 | 110 | |
NM_070459.5 | NP_502860.2 | 4 - 5 | 964 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
