Assay Details
| Gene Symbol: rac-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047496.1 | NP_001040961.1 | 2 - 3 | 196 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rac-2| Gene Symbol: rac-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047496.1 | NP_001040961.1 | 2 - 3 | 196 | 101 | |
Set Membership: |
|