Assay Details
Gene Symbol: sfa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070632.6 | NP_503033.1 | 1 - 2 | 158 | 94 | |
NR_132466.1 | 1 - 2 | 158 | 94 | |||
GenBank mRNA | AJ243905.1 | - | 1 - 2 | 158 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
