Assay Details
| Gene Symbol: itsn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268998.3 | NP_001255927.1 | 7 - 8 | 2748 | 79 | |
| NM_001268999.3 | NP_001255928.1 | 5 - 6 | 1966 | 79 | ||
| NM_001269000.3 | NP_001255929.1 | 4 - 5 | 1387 | 79 | ||
| NM_001313049.4 | NP_001299978.1 | 7 - 8 | 2754 | 79 | ||
| NM_001313050.3 | NP_001299979.1 | 5 - 6 | 1972 | 79 | ||
| NM_001313051.3 | NP_001299980.1 | 4 - 5 | 1393 | 79 | ||
More Information
Set Membership: |
|

itsn-1
Genomic Map