Assay Details
Gene Symbol: egl-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026175.4 | NP_001021346.1 | 10 - 11 | 2270 | 69 | |
NM_001026176.3 | NP_001021347.1 | 10 - 11 | 2270 | 69 | ||
NM_001313085.3 | NP_001300014.1 | 9 - 10 | 2088 | 69 | ||
NM_001313087.3 | NP_001300016.1 | 9 - 10 | 1952 | 69 | ||
NM_001313089.4 | NP_001300018.1 | 9 - 10 | 1959 | 69 | ||
NM_001313095.4 | NP_001300024.1 | 9 | 2139 | 69 | ||
NM_001313097.3 | NP_001300026.1 | 8 - 9 | 2004 | 69 | ||
NM_001313099.3 | NP_001300028.1 | 7 - 8 | 1686 | 69 | ||
NM_001313101.3 | NP_001300030.1 | 7 - 8 | 1686 | 69 | ||
NM_001372980.2 | NP_001360389.1 | 9 - 10 | 1958 | 69 | ||
NM_001380576.2 | NP_001368552.1 | 10 - 11 | 2088 | 69 | ||
NM_001380577.1 | NP_001368553.1 | 9 - 10 | 1958 | 69 | ||
NM_001380580.2 | NP_001368556.1 | 9 - 10 | 2188 | 69 | ||
NM_001380581.2 | NP_001368557.1 | 9 - 10 | 2192 | 69 | ||
NM_001380582.2 | NP_001368558.1 | 8 - 9 | 1856 | 69 | ||
NM_001380583.1 | NP_001368559.1 | 8 - 9 | 1849 | 69 | ||
GenBank mRNA | AF179426.1 | - | 10 - 11 | 2270 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
