Assay Details
Gene Symbol: egl-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026175.4 | NP_001021346.1 | 4 - 5 | 854 | 87 | |
NM_001026176.3 | NP_001021347.1 | 4 - 5 | 854 | 87 | ||
NM_001026177.6 | NP_001021348.1 | 4 - 5 | 860 | 87 | ||
NM_001313085.3 | NP_001300014.1 | 3 | 672 | 87 | ||
NM_001313087.3 | NP_001300016.1 | 4 - 5 | 854 | 87 | ||
NM_001313089.4 | NP_001300018.1 | 5 | 861 | 87 | ||
NM_001313091.3 | NP_001300020.1 | 5 | 860 | 87 | ||
NM_001313093.3 | NP_001300022.1 | 5 | 860 | 87 | ||
NM_001372980.2 | NP_001360389.1 | 5 | 860 | 87 | ||
NM_001380576.2 | NP_001368552.1 | 4 - 5 | 672 | 87 | ||
NM_001380577.1 | NP_001368553.1 | 4 - 5 | 860 | 87 | ||
NM_001380578.2 | NP_001368554.1 | 4 - 5 | 858 | 87 | ||
NM_001380579.1 | NP_001368555.1 | 4 - 5 | 860 | 87 | ||
GenBank mRNA | AF179426.1 | - | 4 - 5 | 854 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
