Assay Details
Gene Symbol: egl-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026175.4 | NP_001021346.1 | 8 - 9 | 1782 | 97 | |
NM_001026176.3 | NP_001021347.1 | 8 - 9 | 1782 | 97 | ||
NM_001313085.3 | NP_001300014.1 | 7 - 8 | 1600 | 97 | ||
NM_001313095.4 | NP_001300024.1 | 7 | 1651 | 97 | ||
NM_001313097.3 | NP_001300026.1 | 6 - 7 | 1516 | 97 | ||
NM_001380576.2 | NP_001368552.1 | 8 - 9 | 1600 | 97 | ||
NM_001380580.2 | NP_001368556.1 | 7 - 8 | 1700 | 97 | ||
NM_001380581.2 | NP_001368557.1 | 7 - 8 | 1704 | 97 | ||
GenBank mRNA | AF179426.1 | - | 8 - 9 | 1782 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
