Assay Details
| Gene Symbol: daam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313190.3 | NP_001300119.1 | 3 - 4 | 326 | 78 | |
| NM_070731.6 | NP_503132.3 | 8 - 9 | 895 | 78 | ||
More Information
Set Membership: |
|

daam-1
Genomic Map