Assay Details
| Gene Symbol: cfap-36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070746.5 | NP_503147.1 | 2 - 3 | 243 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cfap-36| Gene Symbol: cfap-36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070746.5 | NP_503147.1 | 2 - 3 | 243 | 137 | |
Set Membership: |
|