Assay Details
Gene Symbol: tre-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306844.3 | NP_001293773.1 | 8 - 9 | 1079 | 123 | |
NM_001380293.1 | NP_001368400.1 | 7 - 8 | 938 | 123 | ||
NM_068657.7 | NP_501058.2 | 10 - 11 | 1337 | 123 | ||
GenBank mRNA | AJ512334.1 | - | 9 - 10 | 1314 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
