Assay Details
| Gene Symbol: rpn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068663.5 | NP_501064.1 | 5 - 6 | 2720 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpn-1| Gene Symbol: rpn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068663.5 | NP_501064.1 | 5 - 6 | 2720 | 72 | |
Set Membership: |
|