Assay Details
Gene Symbol: mltn-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269056.1 | NP_001255985.1 | 2 - 3 | 315 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: mltn-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269056.1 | NP_001255985.1 | 2 - 3 | 315 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() |