Assay Details
Gene Symbol: ckb-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071172.6 | NP_503573.1 | 6 - 7 | 878 | 96 | |
NR_132528.1 | 5 - 6 | 726 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
