Assay Details
Gene Symbol: ckb-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071172.6 | NP_503573.1 | 8 - 9 | 1167 | 73 | |
NR_132528.1 | 7 - 8 | 1015 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
