Assay Details
| Gene Symbol: ifc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071382.7 | NP_503783.1 | 4 - 5 | 1092 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ifc-1| Gene Symbol: ifc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071382.7 | NP_503783.1 | 4 - 5 | 1092 | 72 | |
Set Membership: |
|