Assay Details
| Gene Symbol: chpf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071429.3 | NP_503830.2 | 2 - 3 | 177 | 89 | |
| NR_133615.1 | 2 - 3 | 188 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chpf-2| Gene Symbol: chpf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071429.3 | NP_503830.2 | 2 - 3 | 177 | 89 | |
| NR_133615.1 | 2 - 3 | 188 | 89 | |||
Set Membership: |
|