Assay Details
| Gene Symbol: srab-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071474.1 | NP_503875.1 | 1 - 2 | 224 | 158 | |
| NR_134542.1 | 1 - 2 | 224 | 158 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srab-8| Gene Symbol: srab-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071474.1 | NP_503875.1 | 1 - 2 | 224 | 158 | |
| NR_134542.1 | 1 - 2 | 224 | 158 | |||
Set Membership: |
|