Assay Details
Gene Symbol: grl-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071596.1 | NP_503997.1 | 2 - 3 | 432 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: grl-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071596.1 | NP_503997.1 | 2 - 3 | 432 | 75 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |