Assay Details
Gene Symbol: mtm-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071637.6 | NP_504038.1 | 2 - 3 | 826 | 83 | |
NM_182367.6 | NP_872167.2 | 2 - 3 | 682 | 83 | ||
GenBank mRNA | AY313178.1 | - | 2 - 3 | 824 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
