Assay Details
| Gene Symbol: crn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072075.8 | NP_504476.1 | 1 - 2 | 233 | 107 | |
| GenBank mRNA | AY303576.1 | - | 1 - 2 | 202 | 107 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  Detect Genomic DNA  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 crn-2
crn-2 Genomic Map
Genomic Map Back To Top
 Back To Top