Assay Details
| Gene Symbol: pcm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129372.4 | NP_001122844.1 | 3 - 4 | 416 | 72 | |
| NM_072150.7 | NP_504551.3 | 3 - 4 | 480 | 72 | ||
| GenBank mRNA | U09669.1 | - | 3 - 4 | 363 | 72 | |
More Information
Set Membership: |
|

pcm-1
Genomic Map