Assay Details
| Gene Symbol: acy-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072152.6 | NP_504553.1 | 15 - 16 | 2757 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acy-2| Gene Symbol: acy-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072152.6 | NP_504553.1 | 15 - 16 | 2757 | 143 | |
Set Membership: |
|