Assay Details
| Gene Symbol: pgap-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072166.5 | NP_504567.1 | 2 - 3 | 231 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgap-3| Gene Symbol: pgap-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072166.5 | NP_504567.1 | 2 - 3 | 231 | 115 | |
Set Membership: |
|