Assay Details
| Gene Symbol: cwf-19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072176.6 | NP_504577.1 | 6 - 7 | 765 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cwf-19L1| Gene Symbol: cwf-19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072176.6 | NP_504577.1 | 6 - 7 | 765 | 103 | |
Set Membership: |
|