Assay Details
| Gene Symbol: spe-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356661.4 | NP_001343858.1 | 10 | 1453 | 71 | |
| NM_001356662.3 | NP_001343857.1 | 10 - 11 | 1510 | 71 | ||
More Information
Set Membership: |
|

spe-49
Genomic Map