Assay Details
Gene Symbol: spe-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356661.4 | NP_001343858.1 | 8 | 1216 | 155 | |
NM_001356662.3 | NP_001343857.1 | 8 - 9 | 1273 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
